missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(3-chloro-4-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63290.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15445605

  • £227.90 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(3-chloro-4-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
444.65
MFCD26143358

Specifications

4-Bromo-N-(3-chloro-4-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
5g
444.65
Solid
97%
MFCD26143358
Videos
SDS