missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Bromo-N-(3-chloro-4-methoxyphenyl)-2-fluorobenzenesulfonamide, 97%
Brand: Alfa Aesar™ H65155.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
4-Bromo-N-(3-chloro-4-methoxyphenyl)-2-fluorobenzenesulfonamide | |
394.64 | |
MFCD27975728 |
Specifications
4-Bromo-N-(3-chloro-4-methoxyphenyl)-2-fluorobenzenesulfonamide | |
1g | |
394.64 |
Solid | |
97% | |
MFCD27975728 |