missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Bromo-N-(3-chloro-4-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand: Alfa Aesar™ H63355.06
Code : NEW
Additional Details : Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
4-Bromo-N-(3-chloro-4-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide | |
432.62 | |
MFCD26143361 |
Specifications
4-Bromo-N-(3-chloro-4-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide | |
5g | |
432.62 |
Solid | |
97% | |
MFCD26143361 |