missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Bromo-N-(3-bromophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand: Alfa Aesar™ H62164.06
Code : NEW
Additional Details : Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
4-Bromo-N-(3-bromophenyl)-3-(trifluoromethyl)benzenesulfonamide | |
459.08 | |
MFCD26143312 |
Specifications
4-Bromo-N-(3-bromophenyl)-3-(trifluoromethyl)benzenesulfonamide | |
Solid | |
459.08 |
5g | |
97% | |
MFCD26143312 |