missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(3,5-dimethoxyphenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H65029.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459326

  • £62.96 / 1g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(3,5-dimethoxyphenyl)-2-fluorobenzenesulfonamide
390.22
MFCD28098738

Specifications

4-Bromo-N-(3,5-dimethoxyphenyl)-2-fluorobenzenesulfonamide
1g
390.22
Solid
97%
MFCD28098738
Videos
SDS