missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(3,5-dichlorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62595.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15417616

  • £53.11 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(3,5-dichlorophenyl)-3-(trifluoromethyl)benzenesulfonamide
449.07
MFCD26143332

Specifications

4-Bromo-N-(3,5-dichlorophenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
449.07
Solid
97%
MFCD26143332
Videos
SDS