missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(3,4-difluorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63684.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15488446

  • £398.56 / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(3,4-difluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
416.16
MFCD26143382

Specifications

4-Bromo-N-(3,4-difluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
5g
416.16
Solid
97%
MFCD26143382
Videos
SDS