missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Bromo-N-(3,4-dichlorophenyl)-2-fluorobenzenesulfonamide, 97%
Brand: Alfa Aesar™ H64698.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
4-Bromo-N-(3,4-dichlorophenyl)-2-fluorobenzenesulfonamide | |
399.06 | |
MFCD28098729 |
Specifications
4-Bromo-N-(3,4-dichlorophenyl)-2-fluorobenzenesulfonamide | |
1g | |
399.06 |
Solid | |
97% | |
MFCD28098729 |