missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-isopropylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63556.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15478366

  • £319.06 / 5g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-isopropylphenyl)-3-(trifluoromethyl)benzenesulfonamide
422.26
MFCD26143374

Specifications

4-Bromo-N-(2-isopropylphenyl)-3-(trifluoromethyl)benzenesulfonamide
5g
422.26
Solid
97%
MFCD26143374
Videos
SDS