missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62253.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15497206

  • £298.92 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
398.17
MFCD26143318

Specifications

4-Bromo-N-(2-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
97%
398.17
Solid
5g
MFCD26143318
Videos
SDS