missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-fluoro-5-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63468.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15438326

  • £298.92 / 5g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-fluoro-5-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
412.2
MFCD26143368

Specifications

4-Bromo-N-(2-fluoro-5-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
97%
412.2
Solid
5g
MFCD26143368
Videos
SDS