missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-ethylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63850.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15428556

  • £478.06 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-ethylphenyl)-3-(trifluoromethyl)benzenesulfonamide
408.23
MFCD26143389

Specifications

4-Bromo-N-(2-ethylphenyl)-3-(trifluoromethyl)benzenesulfonamide
5g
408.23
Solid
97%
MFCD26143389
Videos
SDS