missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-chlorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62413.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15417416

  • £68.26 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-chlorophenyl)-3-(trifluoromethyl)benzenesulfonamide
414.63
MFCD26143325

Specifications

4-Bromo-N-(2-chlorophenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
414.63
Solid
97%
MFCD26143325
Videos
SDS