missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-chlorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62413.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15427416

  • £341.32 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-chlorophenyl)-3-(trifluoromethyl)benzenesulfonamide
414.63
MFCD26143325

Specifications

4-Bromo-N-(2-chlorophenyl)-3-(trifluoromethyl)benzenesulfonamide
5g
414.63
Solid
97%
MFCD26143325
Videos
SDS