missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-chlorophenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64873.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499216

  • £59.78 / 1g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-chlorophenyl)-2-fluorobenzenesulfonamide
364.61
MFCD27392161

Specifications

4-Bromo-N-(2-chlorophenyl)-2-fluorobenzenesulfonamide
1g
364.61
Solid
97%
MFCD27392161
Videos
SDS