missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Bromo-N-(2-bromo-4-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand: Alfa Aesar™ H63975.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
4-Bromo-N-(2-bromo-4-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide | |
477.07 | |
MFCD28098715 |
Specifications
4-Bromo-N-(2-bromo-4-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide | |
1g | |
477.07 |
Solid | |
97% | |
MFCD28098715 |