missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-bromo-4-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63975.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15438616

  • £373.12 / 5g


Estimated Shipment date
from Supplier 14-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-bromo-4-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
477.07
MFCD28098715

Specifications

4-Bromo-N-(2-bromo-4-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
5g
477.07
Solid
97%
MFCD28098715
Videos
SDS