missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2,5-dichlorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62958.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15468046

  • £498.20 / 5g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2,5-dichlorophenyl)-3-(trifluoromethyl)benzenesulfonamide
449.07
MFCD26960686

Specifications

4-Bromo-N-(2,5-dichlorophenyl)-3-(trifluoromethyl)benzenesulfonamide
5g
449.07
Solid
97%
MFCD26960686
Videos
SDS