missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2,4-dimethylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63264.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15438206

  • £54.70 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2,4-dimethylphenyl)-3-(trifluoromethyl)benzenesulfonamide
408.23
MFCD26143357

Specifications

4-Bromo-N-(2,4-dimethylphenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
408.23
Solid
97%
MFCD26143357
Videos
SDS