missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2,4-dimethylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63264.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15448206

  • £273.48 / 5g


Estimated Shipment date
from Supplier 05-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2,4-dimethylphenyl)-3-(trifluoromethyl)benzenesulfonamide
408.23
MFCD26143357

Specifications

4-Bromo-N-(2,4-dimethylphenyl)-3-(trifluoromethyl)benzenesulfonamide
5g
408.23
Solid
97%
MFCD26143357
Videos
SDS