missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2,4-dibromophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63952.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458596

  • £227.90 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2,4-dibromophenyl)-3-(trifluoromethyl)benzenesulfonamide
537.97
MFCD26143391

Specifications

4-Bromo-N-(2,4-dibromophenyl)-3-(trifluoromethyl)benzenesulfonamide
5g
537.97
Solid
97%
MFCD26143391
Videos
SDS