missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2,3-dimethylphenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64293.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15468826

  • £47.81 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2,3-dimethylphenyl)-2-fluorobenzenesulfonamide
358.23
MFCD28098720

Specifications

4-Bromo-N-(2,3-dimethylphenyl)-2-fluorobenzenesulfonamide
1g
358.23
Solid
97%
MFCD28098720
Videos
SDS