missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-9,9-dimethylfluorene, 98%

Brand:  Alfa Aesar™ H64639.03

Code : NEW

Additional Details : CAS Number : 942615-32-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15461919

  • £40.60 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-9,9-dimethylfluorene
273.17
22540802
942615-32-9
MFCD23099415

Specifications

4-Bromo-9,9-dimethylfluorene
Solid
273.17
22540802
942615-32-9
98%
MFCD23099415
1g
Videos
SDS