missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-5-fluoro-N-(3-fluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61695.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15476706

  • £56.29 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-5-fluoro-N-(3-fluorophenyl)benzenesulfonamide
C12H8BrF2NO2S
348.16
MFCD24849002

Specifications

4-Bromo-5-fluoro-N-(3-fluorophenyl)benzenesulfonamide
C12H8BrF2NO2S
MFCD24849002
97%
348.16
1g
Videos
SDS