missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-5-fluoro-2-methoxyaniline, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
4-Bromo-5-fluoro-2-methoxyaniline is used as a laboratory reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromo-5-fluoro-2-methoxyaniline is used as a laboratory reagent.
Notes
Store in a cool place. Keep the container tightly closed. Incompatible with strong-oxidizing agents, strong acids and strong bases.
4-Bromo-5-fluoro-2-methoxyaniline is used as a laboratory reagent.
Notes
Store in a cool place. Keep the container tightly closed. Incompatible with strong-oxidizing agents, strong acids and strong bases.
Specifications
Quantity | 250 mg |
UN Number | UN2811 |
Formula Weight | 220.04 |
Percent Purity | 96% |
Chemical Name or Material | 4-Bromo-5-fluoro-2-methoxyaniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'