missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-5-fluoro-2-iodotoluene, 98%

Brand:  Alfa Aesar™ H63676.MD

Code : NEW

Additional Details : CAS Number : 870704-15-7 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15490489

  • £12.51 / 250mg


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-5-fluoro-2-iodotoluene
870704-15-7
314.92
MFCD06200719

Specifications

4-Bromo-5-fluoro-2-iodotoluene
1.624
Solid
249° to 250°C (dec.)
Light Sensitive
MFCD06200719
870704-15-7
2.145g/mL
98%
250mg
314.92
Videos
SDS