missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-5-chlorothiophene-2-sulfonyl chloride, ≥95%, Thermo Scientific™

Catalog Number p-2701612
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
166964-35-8
C4HBrCl2O2S2
295.974
MFCD00052094
PABOKJJDABSQCK-UHFFFAOYSA-N
3-bromo-2-chlorothiophene-5-sulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulphonyl chloride, 4-bromo-5-chloro-2-thiophenesulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulphonylchloride, 3-bromo-2-chlorothiophene-5-sulphonyl chloride, 2-thiophenesulfonylchloride, 4-bromo-5-chloro, pubchem5475, 4-bromo-5-chloro-thiophene-2-sulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulfonyl, 4-bromanyl-5-chloranyl-thiophene-2-sulfonyl chloride
2797220
4-bromo-5-chlorothiophene-2-sulfonyl chloride
C1=C(SC(=C1Br)Cl)S(=O)(=O)Cl
This item is not returnable. View return policy
166964-35-8
C4HBrCl2O2S2
295.974
MFCD00052094
PABOKJJDABSQCK-UHFFFAOYSA-N
3-bromo-2-chlorothiophene-5-sulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulphonyl chloride, 4-bromo-5-chloro-2-thiophenesulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulphonylchloride, 3-bromo-2-chlorothiophene-5-sulphonyl chloride, 2-thiophenesulfonylchloride, 4-bromo-5-chloro, pubchem5475, 4-bromo-5-chloro-thiophene-2-sulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulfonyl, 4-bromanyl-5-chloranyl-thiophene-2-sulfonyl chloride
2797220
4-bromo-5-chlorothiophene-2-sulfonyl chloride
C1=C(SC(=C1Br)Cl)S(=O)(=O)Cl
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 295.99
Percent Purity 95%
Chemical Name or Material 4-Bromo-5-chlorothiophene-2-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'