missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-3-(trifluoromethyl)-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62155.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15475545

  • £319.06 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-3-(trifluoromethyl)-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide
448.18
MFCD26143311

Specifications

4-Bromo-3-(trifluoromethyl)-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide
97%
MFCD26143311
Solid
448.18
5g
Videos
SDS