missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-3-(trifluoromethyl)-N-(2,3,4-trifluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63429.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15428306

  • £95.61 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-3-(trifluoromethyl)-N-(2,3,4-trifluorophenyl)benzenesulfonamide
434.15
MFCD26143367

Specifications

4-Bromo-3-(trifluoromethyl)-N-(2,3,4-trifluorophenyl)benzenesulfonamide
97%
MFCD26143367
Solid
434.15
1g
Videos
SDS