missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-3-(trifluoromethyl)-N-(2,3,4-trifluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63429.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15438306

  • £478.06 / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-3-(trifluoromethyl)-N-(2,3,4-trifluorophenyl)benzenesulfonamide
434.15
MFCD26143367

Specifications

4-Bromo-3-(trifluoromethyl)-N-(2,3,4-trifluorophenyl)benzenesulfonamide
97%
MFCD26143367
Solid
434.15
5g
Videos
SDS