missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-3-(trifluoromethyl)benzonitrile, 98%

Brand:  Alfa Aesar™ H61459.03

Code : NEW

Additional Details : CAS Number : 1735-53-1 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15448008

  • £24.38 / 1g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-3-(trifluoromethyl)benzonitrile
250.02
UN3439
1735-53-1
MFCD03095347

Specifications

4-Bromo-3-(trifluoromethyl)benzonitrile
Crystalline Solid
250.02
UN3439
1g
1735-53-1
98%
MFCD03095347
Pale yellow
Videos
SDS