missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylbenzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number 11954591
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
72256-93-0
C7H6BrClO2S
269.54
MFCD03094696
JRNDXUKMWFVEIC-UHFFFAOYSA-N
4-bromo-3-methylbenzene-1-sulfonyl chloride, 4-bromo-3-methylbenzenesulphonyl chloride, 4-bromo-3-methylbenzenesulphonylchloride, 4-bromo-3-methyl-benzenesulfonyl chloride, 6-bromo-m-toluene-3-sulfonyl chloride, pubchem15805, acmc-1bho4, jrndxukmwfveic-uhfffaoysa, 4-bromo-3-methylphenyl chlorosulfone, 4-bromo-3-methylbenzene sulfonyl chloride
2737503
4-bromo-3-methylbenzenesulfonyl chloride
CC1=C(Br)C=CC(=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
72256-93-0
C7H6BrClO2S
269.54
MFCD03094696
JRNDXUKMWFVEIC-UHFFFAOYSA-N
4-bromo-3-methylbenzene-1-sulfonyl chloride, 4-bromo-3-methylbenzenesulphonyl chloride, 4-bromo-3-methylbenzenesulphonylchloride, 4-bromo-3-methyl-benzenesulfonyl chloride, 6-bromo-m-toluene-3-sulfonyl chloride, pubchem15805, acmc-1bho4, jrndxukmwfveic-uhfffaoysa, 4-bromo-3-methylphenyl chlorosulfone, 4-bromo-3-methylbenzene sulfonyl chloride
2737503
4-bromo-3-methylbenzenesulfonyl chloride
CC1=C(Br)C=CC(=C1)S(Cl)(=O)=O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 58°C to 60°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 269.55
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-methylbenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-3-methylbenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'