missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3-methylaniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
250 g
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 79.0°C to 83.0°C |
Color | Brown to White |
Boiling Point | 240.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Linear Formula | BrC6H3(CH3)NH2 |
Quantity | 50 g |
Show More |
Safety and Handling
Product Identifier
- 4-Bromo-3-methylaniline
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'