missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-bromo-3-methyl-1H-pyrazol-5-amine hydrobromide, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 256.93 |
Percent Purity | 97% |
Chemical Name or Material | 4-Bromo-3-methyl-1H-pyrazol-5-amine hydrobromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'