missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorotoluene, 98%, Thermo Scientific™

Catalog Number p-3757052
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
452-74-4
MFCD00040940
SLFNGVGRINFJLK-UHFFFAOYSA-N
4-bromo-3-fluorotoluene, 3-fluoro-4-bromotoluene, benzene, 1-bromo-2-fluoro-4-methyl, 3-fluoro-4-bromo-toluene, 4-bromo-3-fluoro-1-methylbenzene, 1-bromo-2-fluoro-4-methyl-benzene, 3-fluoro-4-bromo-toluone, 1-brom-2-fluor-4-methylbenzol, zlchem 562
573280
1-bromo-2-fluoro-4-methylbenzene
CC1=CC(=C(C=C1)Br)F
This item is not returnable. View return policy
452-74-4
MFCD00040940
SLFNGVGRINFJLK-UHFFFAOYSA-N
4-bromo-3-fluorotoluene, 3-fluoro-4-bromotoluene, benzene, 1-bromo-2-fluoro-4-methyl, 3-fluoro-4-bromo-toluene, 4-bromo-3-fluoro-1-methylbenzene, 1-bromo-2-fluoro-4-methyl-benzene, 3-fluoro-4-bromo-toluone, 1-brom-2-fluor-4-methylbenzol, zlchem 562
573280
1-bromo-2-fluoro-4-methylbenzene
CC1=CC(=C(C=C1)Br)F
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.4900g/mL
Boiling Point 95.0°C (50.0 mmHg)
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H4BrFO
Linear Formula CH3C6H3(Br)F
Refractive Index 1.5290 to 1.5310
Quantity 5 g
Specific Gravity 1.49
Molecular Weight (g/mol) 191.01
Formula Weight 191.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Bromo-3-fluorotoluene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'