missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-3-fluorophenylacetonitrile, 95%

Brand:  Alfa Aesar™ H61205.03

Code : NEW

Additional Details : CAS Number : 499983-13-0 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15477708

  • £244.86 / 1g


Estimated Shipment date
from Supplier 22-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-3-fluorophenylacetonitrile
214.03
UN3439
499983-13-0
MFCD12756199

Specifications

4-Bromo-3-fluorophenylacetonitrile
White
95%
214.03
UN3439
499983-13-0
Solid
1g
MFCD12756199
Videos
SDS