missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-3-fluorobenzonitrile, 98%

Brand:  Alfa Aesar™ H61939.22

Code : NEW

Additional Details : CAS Number : 133059-44-6 Weight : 0.10000kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 15438528

  • £391.14 / 100g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-3-fluorobenzonitrile
200.01
UN3439
133059-44-6
MFCD07782069

Specifications

4-Bromo-3-fluorobenzonitrile
Off-white to yellow
98%
200.01
UN3439
133059-44-6
Solid
100g
MFCD07782069
Videos
SDS