missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-4258804
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
351003-51-5
C6H3BrClFO2S
273.51
IZCXVIACMHTZNA-UHFFFAOYSA-N
4-bromo-3-fluorobenzenesulphonyl chloride, 4-bromo-3-fluorobenzene-1-sulfonyl chloride, 3-fluoro-4-bromobenzenesulfonyl chloride, 4-bromo-3-fluorobenzenesulfonylchloride, 4-bromo-3-fluoro-benzenesulfonyl chloride, buttpark 76\07-90, benzenesulfonyl chloride, 4-bromo-3-fluoro, pubchem2220, acmc-209vnt, 4-bromo-3-fluorobenzene sulfonyl chloride
2778218
4-bromo-3-fluorobenzenesulfonyl chloride
C1=CC(=C(C=C1S(=O)(=O)Cl)F)Br
This item is not returnable. View return policy
351003-51-5
C6H3BrClFO2S
273.51
IZCXVIACMHTZNA-UHFFFAOYSA-N
4-bromo-3-fluorobenzenesulphonyl chloride, 4-bromo-3-fluorobenzene-1-sulfonyl chloride, 3-fluoro-4-bromobenzenesulfonyl chloride, 4-bromo-3-fluorobenzenesulfonylchloride, 4-bromo-3-fluoro-benzenesulfonyl chloride, buttpark 76\07-90, benzenesulfonyl chloride, 4-bromo-3-fluoro, pubchem2220, acmc-209vnt, 4-bromo-3-fluorobenzene sulfonyl chloride
2778218
4-bromo-3-fluorobenzenesulfonyl chloride
C1=CC(=C(C=C1S(=O)(=O)Cl)F)Br
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Boiling Point 130.0°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 273.51
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Bromo-3-fluorobenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-3-fluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P232-Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'