missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-3-fluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide, 95%

Brand:  Alfa Aesar™ H65935.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15430027

  • £66.46 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-3-fluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
348.22
MFCD28098844

Specifications

4-Bromo-3-fluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
95%
348.22
Solid
1g
MFCD28098844
Videos
SDS