missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-(chloromethyl)-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Catalog Number p-3540982
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
915707-66-3
C5H6BrClN2
209.471
MFCD09702416
WHHNKVUVLZLAAM-UHFFFAOYSA-N
4-bromo-3-chloromethyl-1-methyl-1h-pyrazole, 4-bromo-3-chloromethyl-1-methylpyrazole, pubchem23664, 1h-pyrazole,4-bromo-3-chloromethyl-1-methyl
24229714
4-bromo-3-(chloromethyl)-1-methylpyrazole
CN1C=C(C(=N1)CCl)Br
This item is not returnable. View return policy
915707-66-3
C5H6BrClN2
209.471
MFCD09702416
WHHNKVUVLZLAAM-UHFFFAOYSA-N
4-bromo-3-chloromethyl-1-methyl-1h-pyrazole, 4-bromo-3-chloromethyl-1-methylpyrazole, pubchem23664, 1h-pyrazole,4-bromo-3-chloromethyl-1-methyl
24229714
4-bromo-3-(chloromethyl)-1-methylpyrazole
CN1C=C(C(=N1)CCl)Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 209.47
Percent Purity 97%
Chemical Name or Material 4-Bromo-3-(chloromethyl)-1-methyl-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'