missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-3-chlorobenzotrifluoride, 98%

Brand:  Alfa Aesar™ H61978.22

Code : NEW

Additional Details : CAS Number : 402-04-0 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15418578

  • £325.00 / 100g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-3-chlorobenzotrifluoride
402-04-0
259.45
MFCD00673985

Specifications

4-Bromo-3-chlorobenzotrifluoride
100g
98%
MFCD00673985
402-04-0
Liquid
259.45
Videos
SDS