missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3,5-dimethylisoxazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Density | 1.484g/mL |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 176.01 |
Percent Purity | 97% |
Chemical Name or Material | 4-Bromo-3, 5-dimethylisoxazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'