missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-bromo-3,5-dimethyl-1-phenyl-1h-pyrazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Synonym | 4-bromo-3,5-dimethyl-1-phenyl-1h-pyrazole, 1h-pyrazole,4-bromo-3,5-dimethyl-1-phenyl, 1-phenyl-3,5-dimethyl-4-bromo-1h-pyrazole, 1h-pyrazole, 4-bromo-3,5-dimethyl-1-phenyl |
Formula Weight | 251.13 |
Percent Purity | 97% |
Chemical Name or Material | 4-Bromo-3, 5-dimethyl-1-phenyl-1H-pyrazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'