missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dichloropyridine, 95%, Thermo Scientific™

Catalog Number p-7055907
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
343781-45-3
C5H2BrCl2N
226.88
MFCD06797978
LOKLQBZBVPYAQO-UHFFFAOYSA-N
3,5-dichloro-4-bromo-pyridine, pubchem21381, 4-bromo-3,5-dichloropyridin, 3,5-dichloro-4-bromopyridine, pyridine,4-bromo-3,5-dichloro, 4-bromanyl-3,5-bis chloranyl pyridine
10680788
4-bromo-3,5-dichloropyridine
ClC1=CN=CC(Cl)=C1Br
This item is not returnable. View return policy
343781-45-3
C5H2BrCl2N
226.88
MFCD06797978
LOKLQBZBVPYAQO-UHFFFAOYSA-N
3,5-dichloro-4-bromo-pyridine, pubchem21381, 4-bromo-3,5-dichloropyridin, 3,5-dichloro-4-bromopyridine, pyridine,4-bromo-3,5-dichloro, 4-bromanyl-3,5-bis chloranyl pyridine
10680788
4-bromo-3,5-dichloropyridine
ClC1=CN=CC(Cl)=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 75°C to 76°C
Color White
Quantity 250 mg
UN Number UN2811
Beilstein 8760531
Formula Weight 226.88
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Bromo-3,5-dichloropyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'