missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-3,5-dichloropyridine, 95%

Brand:  Alfa Aesar™ H64616.MD

Code : NEW

Additional Details : CAS Number : 343781-45-3 Weight : 0.00025kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 250mg

 View more versions of this product

Product Code. 15451899

  • £10.80 / 250mg


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-3,5-dichloropyridine
226.88
UN2811
343781-45-3
MFCD06797978
8760531

Specifications

4-Bromo-3,5-dichloropyridine
Solid
226.88
UN2811
Off-white
250mg
343781-45-3
95%
MFCD06797978
8760531
75° to 76°C
Videos
SDS