missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethyl)phenol, 97%, Thermo Scientific™

Catalog Number p-4228472
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
50824-04-9
C7H4BrF3O
241.007
PDPGERGWEOJVDC-UHFFFAOYSA-N
4-bromo-2-trifluoromethyl phenol, 4-bromo-2-trifluoromethyl benzenol, 5-bromo-2-hydroxybenzotrifluoride, o-cresol, 4-bromo-alpha,alpha,alpha-trifluoro, 4-bromo-2-trifluoromethyl-phenol, phenol, 4-bromo-2-trifluoromethyl, pubchem18454, acmc-209kqd, bromotrifluoromethylbenzenol, ksc490k9d
142784
4-bromo-2-(trifluoromethyl)phenol
C1=CC(=C(C=C1Br)C(F)(F)F)O
This item is not returnable. View return policy
50824-04-9
C7H4BrF3O
241.007
PDPGERGWEOJVDC-UHFFFAOYSA-N
4-bromo-2-trifluoromethyl phenol, 4-bromo-2-trifluoromethyl benzenol, 5-bromo-2-hydroxybenzotrifluoride, o-cresol, 4-bromo-alpha,alpha,alpha-trifluoro, 4-bromo-2-trifluoromethyl-phenol, phenol, 4-bromo-2-trifluoromethyl, pubchem18454, acmc-209kqd, bromotrifluoromethylbenzenol, ksc490k9d
142784
4-bromo-2-(trifluoromethyl)phenol
C1=CC(=C(C=C1Br)C(F)(F)F)O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 85.8°C to 87.1°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 241.01
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)phenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-(trifluoromethyl)phenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'