missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-(trifluoromethyl)benzaldehyde, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Bromo-2-(trifluoromethyl)benzaldehyde is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical intermediate and intermediate in organic syntheses.
Solubility
Sparingly soluble in water (0.083 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.
4-Bromo-2-(trifluoromethyl)benzaldehyde is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical intermediate and intermediate in organic syntheses.
Solubility
Sparingly soluble in water (0.083 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.
Specifications
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Solubility Information | Sparingly soluble in water (0.083g/L) (25°C). |
Formula Weight | 253.02 |
Percent Purity | 95% |
Chemical Name or Material | 4-Bromo-2-(trifluoromethyl)benzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'