4-Bromo-2-thiophenecarbonyl chloride, 97%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: CC28502CB

250MG 4-Bromo-2-thiophenecarbonyl chloride, 95%

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 58777-65-4 Weight: 0.05025kg Transport: UN number : 2923 Chem class : 1a Pack group : III


Packaging Amber Glass Bottle
Quantity 250mg

Product Code. 10029331

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-thiophenecarbonyl chloride
CAS 58777-65-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 95%
Molecular Formula C5H2BrClOS
Formula Weight 225.49
MDL Number MFCD04115391

Specification

Formula Weight 225.49
Quantity 250mg
Chemical Name or Material 4-Bromo-2-thiophenecarbonyl chloride
Assay 97%
Molecular Formula C5H2BrClOS
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Assay Percent Range 95%
MDL Number MFCD04115391
CAS 58777-65-4
CAS Min % 96.5
CAS Max % 100.0
Alternative Products
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.