missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitrotoluene, 98%, Thermo Scientific™

Catalog Number p-3777496
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
60956-26-5
C7H6BrNO2
216.03
MFCD00041243
KZNXALJXBRSMFL-UHFFFAOYSA-N
4-bromo-2-nitrotoluene, 2-nitro-4-bromotoluene, 4-bromo-6-nitrotoluene, bromo 4--2-nitrotoluene, 5-bromo-2-methylnitrobenzene, benzene, 4-bromo-1-methyl-2-nitro, 4-bromo-1-methyl-2-nitro-benzene, 4-brom-2-nitrotoluol, pubchem10403, maybridge1_002072
123546
4-bromo-1-methyl-2-nitrobenzene
CC1=C(C=C(C=C1)Br)[N+](=O)[O-]
This item is not returnable. View return policy
60956-26-5
C7H6BrNO2
216.03
MFCD00041243
KZNXALJXBRSMFL-UHFFFAOYSA-N
4-bromo-2-nitrotoluene, 2-nitro-4-bromotoluene, 4-bromo-6-nitrotoluene, bromo 4--2-nitrotoluene, 5-bromo-2-methylnitrobenzene, benzene, 4-bromo-1-methyl-2-nitro, 4-bromo-1-methyl-2-nitro-benzene, 4-brom-2-nitrotoluol, pubchem10403, maybridge1_002072
123546
4-bromo-1-methyl-2-nitrobenzene
CC1=C(C=C(C=C1)Br)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 43.0°C to 46.0°C
Color Orange to Yellow
Boiling Point 130.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 216.03
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromo-2-nitrotoluene
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-nitrotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'