missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylphenyl isocyanate, 98%, Thermo Scientific™

Catalog Number p-7058362
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1591-98-6
C8H6BrNO
212.046
MFCD00037039
NXAYRHQYINQHRF-UHFFFAOYSA-N
4-bromo-2-methylphenyl isocyanate, benzene,4-bromo-1-isocyanato-2-methyl, 4-bromo-2-methylphenylisocyanate, 4-bromo-2-methylbenzenisocyanate, acmc-1bpqh, 5-bromo-2-isocyanatotoluene, 2-methyl-4-bromophenyl isocyanate, 4-bromanyl-1-isocyanato-2-methyl-benzene
2735594
4-bromo-1-isocyanato-2-methylbenzene
CC1=C(C=CC(=C1)Br)N=C=O
This item is not returnable. View return policy
1591-98-6
C8H6BrNO
212.046
MFCD00037039
NXAYRHQYINQHRF-UHFFFAOYSA-N
4-bromo-2-methylphenyl isocyanate, benzene,4-bromo-1-isocyanato-2-methyl, 4-bromo-2-methylphenylisocyanate, 4-bromo-2-methylbenzenisocyanate, acmc-1bpqh, 5-bromo-2-isocyanatotoluene, 2-methyl-4-bromophenyl isocyanate, 4-bromanyl-1-isocyanato-2-methyl-benzene
2735594
4-bromo-1-isocyanato-2-methylbenzene
CC1=C(C=CC(=C1)Br)N=C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 47°C to 51°C
Flash Point >110°C (230°F)
Odor Irritating
Quantity 1 g
UN Number UN2811
Beilstein 7633645
Sensitivity Moisture sensitive
Formula Weight 212.05
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-methylphenyl isocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'